Assay Details
Target Gene Details
Entrez Gene ID: | 74478 |
Gene Name: | sorting nexin 29 |
Gene Aliases: |
4933437K13Rik, ENSMUSG00000079738, Gm11170, Gm1737, Gm930, Rundc2a |
Location: |
Chr.16:11321256-11755931 on Build GRCm38 |
Assay Gene Location: | Within Exon 22 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Snx29 | NM_001290148.1 | 14 | 1482 | NP_001277077.1 |
NM_028964.4 | 21 | 2531 | NP_083240.2 | |
AF399755.1 | AAK94018.1 | |||
AK017102.1 | 14 | 1483 | BAB30596.1 | |
CR521963.1 | 1 | 147 |
More Information
Set Membership: |
Intragenic Exonic |