Assay Details
Target Gene Details
Entrez Gene ID: | 19280 |
Gene Name: | protein tyrosine phosphatase, receptor type, S |
Gene Aliases: |
AL022616, PTP, PTP-NU3, PTPNU-3, PTPsigma, Ptpt9, R-PTP-S, RPTPsigma |
Location: |
Chr.17:56412426-56476480 on Build GRCm38 |
Assay Gene Location: | Within Exon 8 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ptprs | NM_001252453.1 | 7 | 1138 | NP_001239382.1 |
NM_001252455.1 | 7 | 1138 | NP_001239384.1 | |
NM_001252456.1 | 7 | 1115 | NP_001239385.1 | |
NM_011218.2 | 7 | 1138 | NP_035348.2 | |
BC052462.1 | 7 | 1064 | AAH52462.1 | |
BC083188.1 | 7 | 1097 | AAH83188.1 | |
D28530.1 | 7 | 1027 | BAA05886.1 | |
D28531.1 | 7 | 1027 | BAA05887.1 | |
X82288.1 | 6 | 947 | CAA57732.1 |