Assay Details
Target Gene Details
Entrez Gene ID: | 19280 |
Gene Name: | protein tyrosine phosphatase, receptor type, S |
Gene Aliases: |
AL022616, PTP, PTP-NU3, PTPNU-3, PTPsigma, Ptpt9, R-PTP-S, RPTPsigma |
Location: |
Chr.17:56412426-56476480 on Build GRCm38 |
Assay Gene Location: | Within Exon 28 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ptprs | NM_001252453.1 | 23 | 3769 | NP_001239382.1 |
NM_001252455.1 | 22 | 3757 | NP_001239384.1 | |
NM_001252456.1 | 23 | 3746 | NP_001239385.1 | |
NM_011218.2 | 27 | 4987 | NP_035348.2 | |
AK144123.1 | 15 | 2348 | BAE25714.1 | |
AK159320.1 | 10 | 1050 | BAE34987.1 | |
AK164645.1 | 3 | 371 | BAE37859.1 | |
AK169714.1 | 6 | 743 | BAE41325.1 | |
BC052462.1 | 23 | 3695 | AAH52462.1 | |
BC083188.1 | 22 | 3716 | AAH83188.1 | |
D28530.1 | 27 | 4867 | BAA05886.1 | |
X82288.1 | 26 | 4796 | CAA57732.1 |
More Information
Set Membership: |
Intragenic Exonic |