Assay Details
Target Gene Details
Entrez Gene ID: | 16004 |
Gene Name: | insulin-like growth factor 2 receptor |
Gene Aliases: |
AI661837, CD222, CI-MPR, M6P/IGF2R, Mpr300 |
Location: |
Chr.17:12682406-12769706 on Build GRCm38 |
Assay Gene Location: | Within Exon 48 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Igf2r | NM_010515.2 | 48 | 7497 | NP_034645.2 |
AK053045.1 | 13 | 2089 | BAC35249.1 | |
AK088699.1 | 23 | 3723 | BAC40512.1 | |
AY644391.1 | 1 | 251 | AAT46419.1 | |
BC031190.1 | 15 | 2503 | AAH31190.1 | |
BC043457.1 | 11 | 1698 | AAH43457.1 | |
BC055018.1 | 11 | 1703 | AAH55018.1 | |
BC150817.1 | 48 | 7408 | ||
BC171973.1 | 48 | 7408 | ||
L19500.1 | 4 | 582 | AAA16037.1 | |
U04710.1 | 48 | 7453 | AAA19568.1 |
More Information
Set Membership: |
Intragenic Exonic |