Assay Details
Target Gene Details
Entrez Gene ID: | 71003 |
Gene Name: | protease, serine 41 |
Gene Aliases: |
4931440B09Rik, Tessp-1, Tessp1 |
Location: |
Chr.17:23836768-23844374 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Prss41 | NM_027644.1 | 5 | 453 | NP_081920.1 |
AB049453.1 | 5 | 453 | BAB68561.1 | |
AK016509.1 | 5 | 431 | ||
AK049297.1 | 5 | 470 | BAC33665.1 | |
BC150797.1 | 5 | 469 |
More Information
Set Membership: |
Intragenic Exonic |