Assay Details
Target Gene Details
Entrez Gene ID: | 56490 |
Gene Name: | zinc finger and BTB domain containing 20 |
Gene Aliases: |
1300017A20Rik, 7330412A13Rik, A930017C21Rik, D16Wsu73e, DPZF, HOF, ODA-8S, Oda8, Zfp288 |
Location: |
Chr.16:42875853-43621944 on Build GRCm38 |
Assay Gene Location: | Within Exon 12 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Zbtb20 | NM_001285805.1 | 11 | 2756 | NP_001272734.1 |
NM_019778.2 | 9 | 2703 | NP_062752.2 | |
NM_181058.1 | 7 | 2481 | NP_851401.1 | |
AB221632.1 | 3 | 2168 | ||
AF185576.1 | 7 | 2526 | AAF03152.1 | |
AF194030.1 | 5 | 2318 | AAF06015.1 | |
AK005028.2 | 7 | 2482 | BAB23755.1 | |
AY028963.1 | 4 | 2285 | AAK27374.1 | |
BC031114.1 | 8 | 2589 | AAH31114.1 | |
BC056446.1 | 11 | 2728 | AAH56446.1 |
More Information
Set Membership: |
Intragenic Exonic |