Assay Details
Target Gene Details
Entrez Gene ID: | 56490 |
Gene Name: | zinc finger and BTB domain containing 20 |
Gene Aliases: |
1300017A20Rik, 7330412A13Rik, A930017C21Rik, D16Wsu73e, DPZF, HOF, ODA-8S, Oda8, Zfp288 |
Location: |
Chr.16:42875853-43621944 on Build GRCm38 |
Assay Gene Location: | Within Exon 11 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Zbtb20 | NM_001285805.1 | 10 | 1856 | NP_001272734.1 |
NM_019778.2 | 8 | 1803 | NP_062752.2 | |
NM_181058.1 | 6 | 1581 | NP_851401.1 | |
AB221632.1 | 2 | 1268 | ||
AF185576.1 | 6 | 1626 | AAF03152.1 | |
AF194030.1 | 4 | 1418 | AAF06015.1 | |
AK005028.2 | 6 | 1582 | BAB23755.1 | |
AK034574.1 | 1 | 455 | ||
AY028963.1 | 3 | 1385 | AAK27374.1 | |
BC031114.1 | 7 | 1689 | AAH31114.1 | |
BC056446.1 | 10 | 1828 | AAH56446.1 |
More Information
Set Membership: |
Intragenic Exonic |