Assay Details
Target Gene Details
Entrez Gene ID: | 20815 |
Gene Name: | serine/arginine-rich protein specific kinase 1 |
Gene Aliases: |
AU017960 |
Location: |
Chr.17:28589590-28622636 on Build GRCm38 |
Assay Gene Location: | Within Exon 16 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Srpk1 | NM_016795.3 | 16 | 2029 | NP_058075.2 |
AB012290.1 | 15 | 1987 | BAA25299.1 | |
AJ224115.1 | 16 | 2023 | CAA11833.1 | |
AK015754.1 | 12 | 1557 | ||
AK151152.1 | 16 | 2092 | BAE30158.1 | |
BC005707.1 | 16 | 1998 | AAH05707.1 | |
BC050761.1 | 16 | 2029 | AAH50761.1 |
More Information
Set Membership: |
Intragenic Exonic |