Assay Details
Target Gene Details
Entrez Gene ID: | 71461 |
Gene Name: | PTK7 protein tyrosine kinase 7 |
Gene Aliases: |
8430404F20Rik, chz, mPTK7/CCK4 |
Location: |
Chr.17:46564451-46630049 on Build GRCm38 |
Assay Gene Location: | Within Exon 20 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ptk7 | NM_175168.4 | 20 | 3732 | NP_780377.1 |
AB055408.1 | 6 | 1231 | BAC53806.1 | |
AF531873.1 | 20 | 3582 | AAQ10071.1 | |
AK018379.1 | 2 | 576 | BAE43251.1 | |
AK053044.1 | 20 | 3665 | BAC35248.1 | |
AK163135.1 | 20 | 3665 | BAE37208.1 | |
BC027800.1 | 9 | 1787 | AAH27800.1 | |
BC076578.1 | 20 | 3732 | AAH76578.1 |
More Information
Set Membership: |
Intragenic Exonic |