Assay Details
Target Gene Details
Entrez Gene ID: | 69101 |
Gene Name: | YdjC homolog (bacterial) |
Gene Aliases: |
1810015A11Rik, 4930521M19Rik |
Location: |
Chr.16:17146882-17148857 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ydjc | NM_026940.4 | 5 | 1072 | NP_081216.1 |
AI642588.1 | ||||
AK007503.1 | 5 | 1061 | BAB25075.1 | |
BC115584.1 | ||||
BC115585.1 |