Assay Details
Target Gene Details
Entrez Gene ID: | 78656 |
Gene Name: | bromodomain containing 8 |
Gene Aliases: |
2610007E11Rik, 4432404P07Rik, SMAP |
Location: |
Chr.18:34598615-34624863 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Brd8 | NM_001289606.1 | 1 | 237 | NP_001276535.1 |
NM_001289607.1 | 1 | 237 | NP_001276536.1 | |
NM_030147.3 | 1 | 237 | NP_084423.2 | |
AK172658.1 | BAE43118.1 | |||
CD548974.1 | 1 | 184 | ||
CD555707.1 | 1 | 237 |
Target Gene Details
Entrez Gene ID: | 19348 |
Gene Name: | kinesin family member 20A |
Gene Aliases: |
AA415432, Rab6kifl |
Location: |
Chr.18:34624624-34633277 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Kif20a | NM_001166407.1 | NP_001159879.1 | ||
Y09632.1 | CAA70845.1 |
More Information
Set Membership: |
Intragenic Exonic Non-exonic |