Assay Details
Target Gene Details
Entrez Gene ID: | 16913 |
Gene Name: | proteasome (prosome, macropain) subunit, beta type 8 (large multifunctional peptidase 7) |
Gene Aliases: |
Lmp-7, Lmp7 |
Location: |
Chr.17:34198195-34201454 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Psmb8 | NM_010724.2 | 6 | 1036 | NP_034854.2 |
AK031801.1 | 6 | 861 | BAC27555.1 | |
AK050627.1 | 6 | 864 | BAE20682.1 | |
AK146182.1 | 6 | 883 | BAE26960.1 | |
AK150674.1 | 6 | 864 | BAE29756.1 | |
AK159971.1 | 6 | 853 | BAE35524.1 | |
AK169753.1 | 4 | 1748 | ||
BC013785.1 | 6 | 860 | AAH13785.1 | |
BC051450.1 | 6 | 867 | AAH51450.1 | |
KJ650228.1 | ||||
U22031.1 | AAA75033.1 | |||
U22032.1 | AAA75034.1 | |||
U22033.1 | AAA75035.1 | |||
U22034.1 | AAA75036.1 | |||
U22035.1 | AAA75037.1 | |||
X64449.1 | 6 | 851 | CAA45779.1 |
More Information
Set Membership: |
Intragenic Exonic |