Assay Details
Target Gene Details
Entrez Gene ID: | 12385 |
Gene Name: | catenin (cadherin associated protein), alpha 1 |
Gene Aliases: |
2010010M04Rik, AA517462, AI988031, Catna1 |
Location: |
Chr.18:35118912-35254775 on Build GRCm38 |
Assay Gene Location: | Within Intron 7 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ctnna1 | NM_009818.1 | NP_033948.1 | ||
AK004623.1 | BAB23418.1 | |||
AK036752.1 | ||||
AK148496.1 | BAE28588.1 | |||
AK149721.1 | ||||
AK149738.1 | ||||
AK159339.1 | BAE35001.1 | |||
AK168635.1 | BAE40495.1 | |||
BC048163.1 | AAH48163.1 | |||
BC068316.1 | AAH68316.1 | |||
BC105576.1 | ||||
D90362.1 | BAA14376.1 | |||
X59990.1 | CAA42607.1 |
Target Gene Details
Entrez Gene ID: | 107065 |
Gene Name: | leucine rich repeat transmembrane neuronal 2 |
Gene Aliases: |
AI851755, BB129880, C630011A14Rik, mKIAA0416 |
Location: |
Chr.18:35209003-35215024 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Lrrtm2 | NM_178005.4 | 2 | 1305 | NP_821072.1 |
AK036155.1 | 2 | 1300 | ||
AK043173.1 | 2 | 1161 | BAC31484.1 | |
AK043784.1 | 2 | 1239 | BAC31653.1 | |
AK044705.1 | 2 | 1307 | BAC32042.1 | |
AK046774.1 | 2 | 1306 | BAC32862.1 | |
AK046818.1 | 2 | 1200 | BAC32883.1 | |
AK049920.1 | 2 | 1200 | BAC33985.1 | |
AK122278.1 | 2 | 1291 | BAC65560.1 | |
AY182027.1 | 1 | 877 | AAO67548.1 |
More Information
Set Membership: |
Intragenic Exonic Intronic Non-exonic |