Assay Details
Target Gene Details
Entrez Gene ID: | 57256 |
Gene Name: | protease, serine 21 |
Gene Aliases: |
1700023E12Rik, TESP5, mT4, testisin |
Location: |
Chr.17:23867873-23873113 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Prss21 | NM_020487.4 | 6 | 817 | NP_065233.2 |
AB033822.1 | 6 | 803 | BAB64263.1 | |
AB059415.1 | 6 | 815 | BAB61788.1 | |
AF176209.2 | 6 | 817 | AAF64407.2 | |
AK006271.1 | 6 | 789 | ||
AY005145.1 | 6 | 794 | AAG02255.1 | |
BC049588.1 | 6 | 831 | AAH49588.1 | |
BC116755.1 | 6 | 815 | ||
BC116757.1 | 6 | 815 |
More Information
Set Membership: |
Intragenic Exonic |