Assay Details
Target Gene Details
Entrez Gene ID: | 22247 |
Gene Name: | uridine monophosphate synthetase |
Gene Aliases: |
1700095D23Rik, AA408257, AL033308, BB164745 |
Location: |
Chr.16:33954784-33967302 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Umps | NM_009471.2 | 6 | 2589 | NP_033497.1 |
AK035821.1 | 6 | 2590 | BAC29199.1 | |
AK035997.1 | 5 | 2437 | ||
AK164487.1 |
More Information
Set Membership: |
Intragenic Exonic |