Assay Details
Target Gene Details
Entrez Gene ID: | 106298 |
Gene Name: | RRN3 RNA polymerase I transcription factor homolog (yeast) |
Gene Aliases: |
AL023001, E130302O19Rik, R75565, TIF-1A, Tif1a |
Location: |
Chr.16:13780699-13814841 on Build GRCm38 |
Assay Gene Location: | Within Exon 18 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Rrn3 | NM_001039521.1 | 18 | 2133 | NP_001034610.1 |
AK080397.1 | 18 | 2091 | ||
AK165676.1 | 18 | 2121 | ||
AK170340.1 | 3 | 448 | BAE41730.1 | |
BC034110.1 | 14 | 1648 | AAH34110.1 | |
BC055781.1 | 5 | 751 | AAH55781.1 | |
BC138774.1 | 18 | 2098 | ||
BC138776.1 | 18 | 2098 |
More Information
Set Membership: |
Intragenic Exonic |