Assay Details
Target Gene Details
Entrez Gene ID: | 79044 |
Gene Name: | mitochondrial ribosomal protein S34 |
Gene Aliases: |
0610007F04Rik, 5330430D13Rik, AV001970, Tce2 |
Location: |
Chr.17:24895120-24896274 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Mrps34 | NM_023260.1 | 3 | 658 | NP_075749.1 |
AK002285.1 | 3 | 632 | BAB21988.1 | |
AK004049.1 | 3 | 622 | BAB23142.1 | |
AK007337.1 | 3 | 622 | BAB24970.1 | |
AK019922.1 | 1 | 879 | ||
AK181947.1 | 1 | 45 | ||
AK216319.1 | ||||
BC024336.1 | 3 | 617 | AAH24336.1 |
More Information
Set Membership: |
Intragenic Exonic |