Assay Details
Target Gene Details
Entrez Gene ID: | 224727 |
Gene Name: | BCL2-associated athanogene 6 |
Gene Aliases: |
2410045D21Rik, AA408914, BAG-6, Bat3, D17H6S52E, G3, Scythe |
Location: |
Chr.17:35134600-35147322 on Build GRCm38 |
Assay Gene Location: | Within Exon 11 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Bag6 | NM_001252468.1 | 9 | 1479 | NP_001239397.1 |
NM_001252469.1 | 9 | 1285 | NP_001239398.1 | |
NM_057171.2 | 10 | 1533 | NP_476512.1 | |
AK148796.1 | 9 | 1399 | BAE28666.1 | |
AK164266.1 | 9 | 1256 | ||
AK170134.1 | 10 | 1353 | BAE41587.1 | |
AK190556.1 | ||||
AK193836.1 | ||||
AK198776.1 | 2 | 156 | ||
AK199983.1 | ||||
AK200418.1 | ||||
BC026647.1 | 2 | 277 | AAH26647.1 | |
BC037051.1 | 10 | 1357 | AAH37051.1 |
More Information
Set Membership: |
Intragenic Exonic |