Assay Details
Target Gene Details
Entrez Gene ID: | 22061 |
Gene Name: | transformation related protein 63 |
Gene Aliases: |
AI462811, Ket, P51/P63, P63, P73l, Tp63, Trp53rp1 |
Location: |
Chr.16:25683763-25892103 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Trp63 | NM_001127259.1 | 4 | 978 | NP_001120731.1 |
NM_001127260.1 | 4 | 978 | NP_001120732.1 | |
NM_001127261.1 | 4 | 978 | NP_001120733.1 | |
NM_001127262.1 | 2 | 315 | NP_001120734.1 | |
NM_001127263.1 | 2 | 315 | NP_001120735.1 | |
NM_001127264.1 | 2 | 315 | NP_001120736.1 | |
NM_001127265.1 | 2 | 315 | NP_001120737.1 | |
NM_011641.2 | 2 | 315 | NP_035771.1 | |
AB010152.1 | 2 | 289 | BAA32432.1 | |
AF075434.1 | 4 | 453 | AAC62639.1 | |
AF075435.1 | 4 | 453 | AAC62640.1 | |
AF075436.1 | 4 | 453 | AAC62641.1 | |
AF075437.1 | 2 | 171 | AAC62642.1 | |
AF075438.1 | 2 | 171 | AAC62643.1 | |
AF075439.1 | 2 | 171 | AAC62644.1 | |
AK048623.1 | 2 | 315 | BAC33397.1 | |
AK137253.1 | 2 | 316 | BAE23286.1 | |
AK138305.1 | 4 | 982 | ||
BC090649.1 | 2 | 297 | AAH90649.1 | |
BC092537.1 | 2 | 315 | AAH92537.1 |
More Information
Set Membership: |
Intragenic Exonic |