Assay Details
Target Gene Details
Entrez Gene ID: | 13653 |
Gene Name: | early growth response 1 |
Gene Aliases: |
A530045N19Rik, ETR103, Egr-1, Krox-1, Krox-24, Krox24, NGF1-A, NGFI-A, NGFIA, TIS8, Zenk, Zfp-6, Zif268, egr |
Location: |
Chr.18:34861207-34864956 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Egr1 | NM_007913.5 | 1 | 389 | NP_031939.1 |
AK037837.1 | 2 | 366 | BAC29885.1 | |
AK040925.1 | 1 | 389 | BAC30748.1 | |
AK145198.1 | 1 | 388 | BAE26289.1 | |
AK146322.1 | 1 | 389 | BAE27077.1 | |
BC138613.1 | 1 | 178 | ||
BC138615.1 | 1 | 178 | ||
M19643.1 | AAA39382.1 | |||
M20157.1 | 1 | 365 | AAA37544.1 | |
M38174.1 | AAA39383.1 |
More Information
Set Membership: |
Intragenic Exonic |