Assay Details
Target Gene Details
Entrez Gene ID: | 225372 |
Gene Name: | amyloid beta (A4) precursor protein-binding, family B, member 3 |
Gene Aliases: |
Fe65l2, Rirl2, TR2S |
Location: |
Chr.18:36671154-36679366 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Apbb3 | NM_146085.1 | 1 | 67 | NP_666197.1 |
AK171634.1 | ||||
BC024809.1 | 1 | 67 | AAH24809.1 |
Target Gene Details
Entrez Gene ID: | 67843 |
Gene Name: | solute carrier family 35, member A4 |
Gene Aliases: |
2610030J16Rik |
Location: |
Chr.18:36679215-36683862 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Slc35a4 | NM_001083317.1 | 1 | 86 | NP_001076786.1 |
NM_026404.2 | 1 | 86 | NP_080680.2 | |
BY088803.1 | 1 | 86 | ||
BY732053.1 | 1 | 65 |
More Information
Set Membership: |
Intragenic Exonic |