Assay Details
Target Gene Details
Entrez Gene ID: | 268930 |
Gene Name: | protein kinase, membrane associated tyrosine/threonine 1 |
Gene Aliases: |
6230424P17, AW209059, Myt1 |
Location: |
Chr.17:23726336-23736735 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Pkmyt1 | NM_023058.3 | 2 | 628 | NP_075545.2 |
AF175892.1 | 2 | 513 | AAG09210.1 | |
AK031787.1 | 2 | 496 | ||
AK158550.1 | 2 | 629 | BAE34554.1 | |
AK165164.1 | 2 | 495 | BAE38056.1 | |
BC025061.1 | 2 | 351 | AAH25061.1 |
More Information
Set Membership: |
Intragenic Exonic |