Assay Details
Target Gene Details
Entrez Gene ID: | 17256 |
Gene Name: | male enhanced antigen 1 |
Gene Aliases: |
Mea-1 |
Location: |
Chr.17:46679138-46683126 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Mea1 | NM_001277309.1 | 2 | 287 | NP_001264238.1 |
NM_001277310.1 | 2 | 189 | NP_001264239.1 | |
NM_001277311.1 | 2 | 205 | NP_001264240.1 | |
NM_001277312.1 | 2 | 232 | NP_001264241.1 | |
NM_010787.2 | 2 | 224 | NP_034917.1 | |
NR_102392.1 | 2 | 316 | ||
AK005106.1 | 2 | 173 | BAB23822.1 | |
AK005369.1 | 2 | 177 | BAB23978.1 | |
AK008194.1 | 2 | 183 | BAB25525.1 | |
AK008305.1 | 2 | 185 | BAB25590.1 | |
AK010863.1 | 2 | 194 | BAB27231.1 | |
AK010904.1 | 2 | 222 | BAB27257.1 | |
AK010962.1 | 2 | 179 | BAB27294.1 | |
AK011189.1 | 2 | 166 | BAB27454.1 | |
AK012617.1 | 2 | 179 | BAB28358.1 | |
BC013344.1 | 1 | 187 | AAH13344.1 | |
BG085669.2 | 2 | 184 | ||
BY707372.1 | 2 | 270 | ||
CJ042312.1 | 2 | 275 | ||
D17341.1 | 2 | 163 | BAA04159.1 | |
L10401.1 | 2 | 208 | AAA39520.1 | |
M27938.1 | 2 | 208 | AAA39519.1 |