Assay Details
Target Gene Details
Entrez Gene ID: | 13078 |
Gene Name: | cytochrome P450, family 1, subfamily b, polypeptide 1 |
Gene Aliases: |
CP1B, P4501b1 |
Location: |
Chr.17:79696282-79715065 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Cyp1b1 | NM_009994.1 | 3 | 4509 | NP_034124.1 |
AK016699.1 | ||||
AK139368.1 | 4 | 2721 | BAE23978.1 | |
U03283.1 | 3 | 4509 | AAC52141.1 |
More Information
Set Membership: |
Intragenic Exonic |