Assay Details
Target Gene Details
Entrez Gene ID: | 21815 |
Gene Name: | TGFB-induced factor homeobox 1 |
Gene Aliases: |
AA959811, AI462167, Tgif |
Location: |
Chr.17:70844205-70853532 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Tgif1 | NM_001164074.1 | 3 | 1195 | NP_001157546.1 |
NM_001164075.1 | 3 | 1535 | NP_001157547.1 | |
NM_001164076.1 | 3 | 1261 | NP_001157548.1 | |
NM_001164077.1 | 2 | 1489 | NP_001157549.1 | |
NM_009372.3 | 3 | 1449 | NP_033398.2 | |
AK008909.1 | 3 | 1451 | ||
AK160193.1 | 3 | 1154 | BAE35685.1 | |
BC005724.1 | 3 | 1448 | AAH05724.1 | |
BC012700.1 | 3 | 1546 | AAH12700.1 |
More Information
Set Membership: |
Intragenic Exonic |