Assay Details
Target Gene Details
Entrez Gene ID: | 224902 |
Gene Name: | scaffold attachment factor B2 |
Gene Aliases: |
AA389433, AI255170, mKIAA0138 |
Location: |
Chr.17:56560971-56584672 on Build GRCm38 |
Assay Gene Location: | Within Exon 15 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Safb2 | NM_001029979.2 | 15 | 2329 | NP_001025150.2 |
BC028827.1 | 9 | 1043 | AAH28827.1 | |
BC050855.1 | 15 | 2263 | AAH50855.1 |