Assay Details
Target Gene Details
Entrez Gene ID: | 12053 |
Gene Name: | B cell leukemia/lymphoma 6 |
Gene Aliases: |
Bcl5 |
Location: |
Chr.16:23965052-23988612 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Bcl6 | NM_009744.3 | 5 | 1123 | NP_033874.1 |
AK036975.1 | 5 | 1147 | BAC29654.1 | |
AK039228.1 | 5 | 1145 | BAC30286.1 | |
AK151229.1 | 5 | 1148 | BAE30221.1 | |
BC052315.1 | 5 | 1124 | AAH52315.1 | |
D38377.1 | 5 | 1112 | BAA07456.1 | |
U41465.1 | 4 | 1135 | AAB17432.1 |
More Information
Set Membership: |
Intragenic Exonic |