Assay Details
Target Gene Details
Entrez Gene ID: | 80287 |
Gene Name: | apolipoprotein B mRNA editing enzyme, catalytic polypeptide 3 |
Gene Aliases: |
Arp3, BC003314, Cem15, Rfv3 |
Location: |
Chr.15:79892408-79908905 on Build GRCm38 |
Assay Gene Location: | Within Exon 9 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Apobec3 | NM_001160415.1 | 9 | 2083 | NP_001153887.1 |
NM_030255.3 | 8 | 1984 | NP_084531.2 | |
AA288216.1 | ||||
AK044394.1 | 8 | 2383 | BAC31901.1 | |
AK153719.1 | BAE32154.1 |
More Information
Set Membership: |
Intragenic Exonic |