Assay Details
Target Gene Details
Entrez Gene ID: | 13972 |
Gene Name: | guanine nucleotide binding protein (G protein), beta polypeptide 1-like |
Gene Aliases: |
ESTM55, Gm16314, Me49f07, OTTMUSG00000033458, Wdr14, Wdvcf |
Location: |
Chr.16:18498021-18566680 on Build GRCm38 |
Assay Gene Location: | Within Exon 11 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Gnb1l | NM_001081682.2 | 8 | 2291 | NP_001075151.1 |
NM_001285491.1 | 8 | 2287 | NP_001272420.1 | |
NM_001285493.1 | 9 | 2326 | NP_001272422.1 | |
NM_001285494.1 | 8 | 2191 | NP_001272423.1 | |
NM_023120.5 | 8 | 2233 | NP_075609.2 | |
NR_104347.1 | 9 | 2358 | ||
AK158210.1 | 9 | 2313 | BAE34406.1 | |
AK166195.1 | 8 | 2175 | BAE38622.1 | |
AK173227.1 | 8 | 2216 | BAD32505.1 |
More Information
Set Membership: |
Intragenic Exonic |