Assay Details
Target Gene Details
Entrez Gene ID: | 16419 |
Gene Name: | integrin beta 5 |
Gene Aliases: |
AA475909, AI874634, ESTM23, [b]-5, [b]5, [b]5A, [b]5B, beta-5, beta5 |
Location: |
Chr.16:33829665-33949338 on Build GRCm38 |
Assay Gene Location: | Within Exon 10 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Itgb5 | NM_001145884.1 | 10 | 1699 | NP_001139356.1 |
NM_010580.2 | 10 | 1699 | NP_034710.2 | |
AF022110.1 | 10 | 1698 | AAD08782.1 | |
AF043256.1 | 10 | 1604 | AAC40109.1 | |
AF043257.1 | 10 | 1604 | AAC40110.1 | |
AK087476.1 | 10 | 1445 | ||
AK140251.1 | 10 | 1611 | BAE24299.1 | |
AK143532.1 | 10 | 1634 | ||
BC046280.1 | 10 | 1608 | ||
BC058246.1 | 10 | 1663 | AAH58246.1 |
More Information
Set Membership: |
Intragenic Exonic |