Assay Details
Target Gene Details
Entrez Gene ID: | 218772 |
Gene Name: | retinoic acid receptor, beta |
Gene Aliases: |
A830025K23, Hap, Nr1b2 |
Location: |
Chr.14:16430839-17082331 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Rarb | NM_001289760.1 | NP_001276689.1 | ||
NM_001289761.1 | NP_001276690.1 | |||
NM_001289762.1 | NP_001276691.1 | |||
NM_011243.2 | 1 | 326 | NP_035373.1 | |
AK043726.1 | 1 | 679 | ||
AK054178.1 | 1 | 290 | ||
AK078471.1 | 1 | 39 | ||
AK190447.1 | ||||
BC076597.1 | 1 | 322 | AAH76597.1 | |
S56660.1 | 1 | 222 | AAB25784.2 | |
X56569.1 | CAA39918.1 | |||
X56573.1 | 1 | 446 | CAA39920.1 |
More Information
Set Membership: |
Intragenic Exonic |