Assay Details
Target Gene Details
Entrez Gene ID: | 16520 |
Gene Name: | potassium inwardly-rectifying channel, subfamily J, member 4 |
Gene Aliases: |
IRK3, Kcnf2, MB-IRK3 |
Location: |
Chr.15:79483714-79511883 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Kcnj4 | NM_008427.4 | 2 | 1450 | NP_032453.3 |
AB024304.1 | BAA75518.1 | |||
AI848768.1 | 1 | 102 | ||
BC120593.1 | 2 | 1427 | ||
BC137594.1 | 2 | 1427 | ||
S71382.1 | 2 | 1451 | AAB31087.1 | |
U11075.1 | 2 | 1420 | AAA53241.1 |
More Information
Set Membership: |
Intragenic Exonic |