Assay Details
Target Gene Details
Entrez Gene ID: | 22762 |
Gene Name: | zinc finger protein, multitype 2 |
Gene Aliases: |
B330005D23Rik, FOG-2, FOG2 |
Location: |
Chr.15:40655042-41104592 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Zfpm2 | NM_011766.5 | 1 | 274 | NP_035896.1 |
AF118845.1 | 1 | 44 | AAD17251.1 | |
AF125166.1 | 1 | 275 | AAD33250.1 | |
AK033131.1 | BAC28166.1 | |||
AK046521.1 | 1 | 282 | ||
BC059241.1 | 1 | 174 | AAH59241.1 |
More Information
Set Membership: |
Intragenic Exonic |