Assay Details
Target Gene Details
Entrez Gene ID: | 239134 |
Gene Name: | guanylate cyclase 1, soluble, beta 2 |
Gene Aliases: |
B930025B03, GC-SB2 |
Location: |
Chr.14:62373023-62457562 on Build GRCm38 |
Assay Gene Location: | Within Exon 17 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Gucy1b2 | NM_001204340.1 | 17 | 2572 | NP_001191269.1 |
NM_172810.3 | 17 | 2617 | NP_766398.1 | |
AK047129.1 | 17 | 2618 | BAC32968.1 | |
BC137868.1 | ||||
BC144988.1 | ||||
BY391897.1 | 2 | 312 |
More Information
Set Membership: |
Intragenic Exonic |