Assay Details
Target Gene Details
Entrez Gene ID: | 105787 |
Gene Name: | protein kinase, AMP-activated, alpha 1 catalytic subunit |
Gene Aliases: |
AI194361, AI450832, AL024255, AMPKalpha1, C130083N04Rik |
Location: |
Chr.15:5143861-5181899 on Build GRCm38 |
Assay Gene Location: | Within Exon 9 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Prkaa1 | NM_001013367.3 | 9 | 3577 | NP_001013385.3 |
AK081874.1 | 3 | 2432 | BAC38355.1 | |
AK147811.1 | 9 | 3555 | ||
AK160612.1 | 9 | 3558 | BAE35914.1 | |
BC086695.1 | 9 | 3547 |
More Information
Set Membership: |
Intragenic Exonic |