Assay Details
Target Gene Details
Entrez Gene ID: | 106014 |
Gene Name: | family with sequence similarity 19, member A5 |
Gene Aliases: |
TAFA5, Tafa-5, Tara-5 |
Location: |
Chr.15:87544299-87759364 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Fam19a5 | NM_001252310.1 | NP_001239239.1 | ||
NM_134096.2 | 1 | 120 | NP_598857.1 | |
AK090194.1 | BAC41130.1 | |||
AK090307.1 | ||||
AY325124.1 | 1 | 121 | AAP92416.1 | |
BC015306.1 | 1 | 121 | AAH15306.1 | |
BU057253.1 | ||||
BY733542.1 |
More Information
Set Membership: |
Intragenic Exonic |