Assay Details
Target Gene Details
Entrez Gene ID: | 12563 |
Gene Name: | cadherin 6 |
Gene Aliases: |
K-cadherin, cad6, cadherin-6 |
Location: |
Chr.15:13028699-13173689 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Cdh6 | NM_007666.3 | 3 | 738 | NP_031692.2 |
AK134611.1 | 3 | 775 | BAE22207.1 | |
BC107008.1 | 2 | 479 | AAI07009.1 | |
BC107013.1 | 2 | 479 | AAI07014.1 | |
D82029.1 | 2 | 479 | BAA11516.1 |
More Information
Set Membership: |
Intragenic Exonic |