Assay Details
Target Gene Details
Entrez Gene ID: | 76355 |
Gene Name: | TDP-glucose 4,6-dehydratase |
Gene Aliases: |
2610017J16Rik, 2610025M23Rik, AI648925 |
Location: |
Chr.14:118111911-118132765 on Build GRCm38 |
Assay Gene Location: | Within Exon 12 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Tgds | NM_029578.3 | 12 | 1445 | NP_083854.3 |
AK011439.1 | 11 | 1333 | ||
AK011555.1 | 12 | 1338 | BAB27693.1 | |
AK154341.1 | 12 | 1422 | BAE32527.1 | |
AK165685.1 | 11 | 1328 | ||
AK165955.1 | 12 | 1429 | BAE38483.1 | |
BC021419.1 | 12 | 1421 | AAH21419.1 |
More Information
Set Membership: |
Intragenic Exonic |