Assay Details
Target Gene Details
Entrez Gene ID: | 105651 |
Gene Name: | protein phosphatase 1, regulatory (inhibitor) subunit 3E |
Gene Aliases: |
A630071A11Rik, AW049239, Gm1211 |
Location: |
Chr.14:54875597-54877538 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ppp1r3e | NM_001167908.1 | 2 | 1253 | NP_001161380.1 |
AK044095.1 | 1 | 426 | BAC31772.1 |
More Information
Set Membership: |
Intragenic Exonic |