Assay Details
Target Gene Details
Entrez Gene ID: | 26942 |
Gene Name: | sperm associated antigen 1 |
Gene Aliases: |
tpis |
Location: |
Chr.15:36175483-36235621 on Build GRCm38 |
Assay Gene Location: | Within Exon 19 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Spag1 | NM_012031.3 | 19 | 2946 | NP_036161.2 |
AF181252.1 | 19 | 2938 | AAF06160.1 | |
AF181253.1 | 9 | 1630 | AAF06161.1 | |
AK032601.1 | 16 | 2741 | BAC27944.1 | |
BC046313.1 | 19 | 2946 | AAH46313.1 |