Assay Details
Target Gene Details
Entrez Gene ID: | 215654 |
Gene Name: | cadherin 12 |
Gene Aliases: |
Cdhb |
Location: |
Chr.15:20448879-21597005 on Build GRCm38 |
Assay Gene Location: | Within Exon 11 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Cdh12 | NM_001008420.2 | 11 | 1964 | NP_001008420.1 |
BC086655.1 | 11 | 1964 | AAH86655.1 |
More Information
Set Membership: |
Intragenic Exonic |