Assay Details
Target Gene Details
Entrez Gene ID: | 66168 |
Gene Name: | glutamate receptor, ionotropic, N-methyl D-aspartate-associated protein 1 (glutamate binding) |
Gene Aliases: |
1110025J15Rik, Lag, Tmbim3 |
Location: |
Chr.15:76246740-76249911 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Grina | NM_023168.3 | 2 | 194 | NP_075657.1 |
AF182040.1 | 2 | 177 | AAG16897.1 | |
AK003920.1 | 2 | 203 | ||
BC010802.1 | 2 | 227 | AAH10802.1 | |
BC019157.1 | 2 | 215 | AAH19157.1 | |
BC037667.1 | 2 | 194 | AAH37667.2 |