Assay Details
Target Gene Details
Entrez Gene ID: | 76485 |
Gene Name: | glycosyltransferase 8 domain containing 1 |
Gene Aliases: |
2410004H05Rik, 5430414N14Rik, AI450005 |
Location: |
Chr.14:31001347-31012441 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 1 - Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Glt8d1 | NM_001165930.1 | NP_001159402.1 | ||
NM_029626.2 | NP_083902.2 | |||
AK077388.1 | ||||
CJ177766.1 |
Target Gene Details
Entrez Gene ID: | 69019 |
Gene Name: | signal peptidase complex subunit 1 homolog (S. cerevisiae) |
Gene Aliases: |
1810004F21Rik |
Location: |
Chr.14:30999826-31001666 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Spcs1 | NM_026911.3 | 1 | 230 | NP_081187.2 |
BX512708.1 | 1 | 172 | ||
CX205717.1 |
More Information
Set Membership: |
Intragenic Exonic |