Assay Details
Target Gene Details
Entrez Gene ID: | 12469 |
Gene Name: | chaperonin containing Tcp1, subunit 8 (theta) |
Gene Aliases: |
AI132397, Cctq, Tcpq |
Location: |
Chr.16:87483325-87495869 on Build GRCm38 |
Assay Gene Location: | Within Exon 15 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Cct8 | NM_009840.3 | 15 | 1781 | NP_033970.3 |
AK014005.1 | 8 | 847 | BAB29110.1 | |
AK075711.1 | 15 | 1762 | ||
AK075880.1 | 15 | 1770 | BAC36025.1 | |
AK145661.1 | 15 | 1765 | BAE26573.1 | |
AK145754.1 | 15 | 1764 | BAE26629.1 | |
AK145918.1 | 15 | 1783 | BAE26749.1 | |
AK147038.1 | 15 | 1784 | BAE27627.1 | |
AK159969.1 | 15 | 1772 | ||
AK168465.1 | 15 | 1755 | ||
AK172867.1 | 15 | 1752 | BAD32145.1 | |
BC009007.1 | 15 | 1739 | AAH09007.1 | |
Z37164.1 | 15 | 1785 | CAA85521.1 |
More Information
Set Membership: |
Intragenic Exonic |