Assay Details
Target Gene Details
Entrez Gene ID: | 239510 |
Gene Name: | PHD finger protein 20-like 1 |
Gene Aliases: |
CGI-72, E130113K22Rik |
Location: |
Chr.15:66577517-66647043 on Build GRCm38 |
Assay Gene Location: | Within Exon 21 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Phf20l1 | NM_001081409.1 | 21 | 3787 | NP_001074878.1 |
AK046670.1 | 4 | 1307 | BAC32830.1 | |
AK053606.1 | 11 | 2146 | BAC35444.1 |