Assay Details
Target Gene Details
Entrez Gene ID: | 30044 |
Gene Name: | opsin 4 (melanopsin) |
Gene Aliases: |
1110007J02Rik, Gm533 |
Location: |
Chr.14:34590618-34600142 on Build GRCm38 |
Assay Gene Location: | Within Exon 10 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Opn4 | NM_001128599.1 | 10 | 1903 | NP_001122071.1 |
AK027928.1 | 2 | 450 | ||
EU303117.1 | 10 | 1719 |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() |
Panther Classification:
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)