Assay Details
Target Gene Details
Entrez Gene ID: | 13840 |
Gene Name: | Eph receptor A6 |
Gene Aliases: |
Ehk2, Hek12, m-ehk2 |
Location: |
Chr.16:59653061-60606484 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Epha6 | NM_007938.2 | 3 | 1286 | NP_031964.2 |
AK090366.1 | 3 | 1287 | ||
BC141090.1 | 3 | 802 | ||
U58332.1 | 3 | 1002 | AAB53836.1 |
More Information
Set Membership: |
Intragenic Exonic |