Assay Details
Target Gene Details
Entrez Gene ID: | 207781 |
Gene Name: | C2 calcium-dependent domain containing 2 |
Gene Aliases: |
5730563M15, 5830404H04Rik, AI314146, AI449533, BF168443, ORF25, mKIAA4045 |
Location: |
Chr.16:97855209-97922651 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
C2cd2 | NM_174847.2 | 1 | 477 | NP_777272.2 |
AK017896.1 | 1 | 452 | ||
AK077731.1 | 1 | 437 | ||
AK080956.1 | 1 | 421 | ||
AK086520.1 | 1 | 478 | ||
AK143875.1 | 1 | 451 | BAE25577.1 | |
AK169902.1 | 1 | 413 | BAE41447.1 | |
AK220454.1 | 1 | 462 | BAD90483.1 | |
BC043688.1 | 1 | 438 | AAH43688.1 |