Assay Details
Target Gene Details
Entrez Gene ID: | 217893 |
Gene Name: | phosphofurin acidic cluster sorting protein 2 |
Gene Aliases: |
6720425G15Rik, AW208651, AW489976, E230011J22, Pacs1l, mKIAA0602 |
Location: |
Chr.12:113014507-113074401 on Build GRCm38 |
Assay Gene Location: | Within Exon 24 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Pacs2 | NM_001081170.1 | 24 | 4518 | NP_001074639.1 |
NM_001291444.1 | 24 | 4524 | NP_001278373.1 | |
NM_001291445.1 | 23 | 4428 | NP_001278374.1 | |
AK122326.1 | 22 | 4207 | BAC65608.1 | |
BC043302.1 | 6 | 2344 | AAH43302.1 | |
BC059076.1 | 10 | 3000 | AAH59076.1 |