Assay Details
Target Gene Details
Entrez Gene ID: | 380768 |
Gene Name: | coiled-coil domain containing 177 |
Gene Aliases: |
Gm1568 |
Location: |
Chr.12:80755447-80760715 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ccdc177 | NM_001008423.2 | 2 | 3720 | NP_001008423.2 |
AK162881.1 | 1 | 500 | ||
BC067044.1 | 2 | 3692 | AAH67044.1 |